Learn More
Ethyl 6-bromohexanoate, 97+%
CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol
Supplier: Thermo Scientific Chemicals A1839818
Description
Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.
Solubility
Soluble in water (water, 64.76 mg/L @ 25°C (est.)).
Notes
Store away from strong oxidizing agents, strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Ethyl 6-bromohexanoate | |
| 1.254 | |
| >110°C (230°F) | |
| 1.4595 | |
| 50 g | |
| 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953 | |
| DXBULVYHTICWKT-UHFFFAOYSA-N | |
| ethyl 6-bromohexanoate | |
| 117544 | |
| ≥97% |
| 25542-62-5 | |
| 128°C to 130°C (16 mmHg) | |
| C8H15BrO2 | |
| MFCD00000270 | |
| 1756051 | |
| Soluble in water (water,64.76mg/L @ 25°C (est.)). | |
| CCOC(=O)CCCCCBr | |
| 223.11 | |
| 223.12 |
Chemical Identifiers
| 25542-62-5 | |
| 223.11 | |
| DXBULVYHTICWKT-UHFFFAOYSA-N | |
| 117544 | |
| CCOC(=O)CCCCCBr |
| C8H15BrO2 | |
| MFCD00000270 | |
| 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953 | |
| ethyl 6-bromohexanoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P374-P380-P501c
H226-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 247-085-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only