missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromohexanoate, 97+%

Catalog No. AAA1839830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1839830 250 g
AAA1839809 10 g
AAA1839818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1839830 Supplier Thermo Scientific Chemicals Supplier No. A1839830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol

Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

Solubility
Soluble in water (water, 64.76 mg/L @ 25°C (est.)).

Notes
Store away from strong oxidizing agents, strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Density 1.254
Boiling Point 128°C to 130°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4595
Quantity 250 g
Beilstein 1756051
Solubility Information Soluble in water (water,64.76mg/L @ 25°C (est.)).
Formula Weight 223.12
Percent Purity ≥97%
Chemical Name or Material Ethyl 6-bromohexanoate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P374-P380-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 247-085-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only