missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 6-Aminopyridine-2-carboxylate 98.0+%, TCI America™
Supplier: TCI America E12135G
Specifications
| Ethyl 6-Aminopyridine-2-carboxylate | |
| 72°C | |
| C8H10N2O2 | |
| 5 g | |
| DTYDENRYZSEGNL-UHFFFAOYSA-N | |
| ethyl 6-aminopyridine-2-carboxylate | |
| 11959065 | |
| ≥98.0% (GC,T) |
| 69142-64-9 | |
| Yellow | |
| MFCD01009842 | |
| 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester | |
| CCOC(=O)C1=NC(=CC=C1)N | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 69142-64-9 | |
| 166.18 | |
| DTYDENRYZSEGNL-UHFFFAOYSA-N | |
| 11959065 | |
| CCOC(=O)C1=NC(=CC=C1)N |
| C8H10N2O2 | |
| MFCD01009842 | |
| 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester | |
| ethyl 6-aminopyridine-2-carboxylate |
Safety and Handling
TSCA : No