CAS RN 69142-64-9
CAS RN 69142-64-9
IUPAC Name:
ethyl 6-aminopyridine-2-carboxylate
Synonyms:
ethyl 6-aminopyridine-2-carboxylate
Molecular Weight (g/mol):
166.18
Molecular Formula:
C8H10N2O2
InChi Key:
DTYDENRYZSEGNL-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NC(N)=CC=C1
1
–
1
of
1
results
1
Ethyl 6-Aminopyridine-2-carboxylate 98.0+%, TCI America™
CAS: 69142-64-9 Molecular Formula: C8H10N2O2 Molecular Weight (g/mol): 166.18 MDL Number: MFCD01009842 InChI Key: DTYDENRYZSEGNL-UHFFFAOYSA-N Synonym: 6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester PubChem CID: 11959065 IUPAC Name: ethyl 6-aminopyridine-2-carboxylate SMILES: CCOC(=O)C1=NC(=CC=C1)N