missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Nitrobenzofuran-2-carboxylate 97.0+%, TCI America™

Catalog No. E09505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09505G 5 g
E095025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09505G Supplier TCI America Supplier No. E09505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69604-00-8
Molecular Formula C11H9NO5
Molecular Weight (g/mol) 235.195
MDL Number MFCD00275272
InChI Key ATHBGWVHAWGMAL-UHFFFAOYSA-N
Synonym ethyl 5-nitrobenzofuran-2-carboxylate, ethyl 5-nitrobenzo b furan-2-carboxylate, 2-benzofurancarboxylic acid, 5-nitro-, ethyl ester, 5-nitrobenzofuran-2-carboxylic acid ethyl ester, ethyl 5-nitrobenzo d furan-2-carboxylate, 5-nitrobenzo b furan-2-carboxylic acid ethyl ester, pubchem15412, 5-nitro-2-benzofurancarboxylic acid ethyl ester, ethylnitrobenzofurancarboxylate, ksc352c3t
PubChem CID 1382954
IUPAC Name ethyl 5-nitro-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(O1)C=CC(=C2)[N+](=O)[O-]

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 235.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Nitrobenzofuran-2-carboxylate