accessibility menu, dialog, popup

UserName

H3COOONOO

CAS RN 69604-00-8

IUPAC Name: ethyl 5-nitro-1-benzofuran-2-carboxylate
Synonyms: ethyl 5-nitro-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 235.2
Molecular Formula: C11H9NO5
InChi Key: ATHBGWVHAWGMAL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(=CC=C2O1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results