missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-nitro-2-furoate, 97%

Catalog No. p-7027826
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2486103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2486103 Supplier Thermo Scientific Chemicals Supplier No. B2486103

CAS: 943-37-3 | C7H7NO5 | 185.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 943-37-3
Molecular Formula C7H7NO5
Molecular Weight (g/mol) 185.135
MDL Number MFCD00092566
InChI Key JMNXLAQKIHVFIC-UHFFFAOYSA-N
PubChem CID 70341
IUPAC Name ethyl 5-nitrofuran-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)[N+](=O)[O-]

Specifications

Melting Point 98°C to 101°C
Quantity 1 g
Synonym ethyl 5-nitro-2-furoate, 2-furoic acid, 5-nitro-, ethyl ester, 5-nitro-furan-2-carboxylic acid ethyl ester, 2-furancarboxylic acid, 5-nitro-, ethyl ester, ethyl5-nitro-2-furoate, acmc-20aky3, ethyl-5-nitro-2-furoate, ethyl 5-nitro-2-furancarboxylate, 5-nitro-2-furoic acid ethyl ester, 5-nitro-2-furancarboxylic acid ethyl ester
Formula Weight 185.14
Percent Purity 97%
Chemical Name or Material Ethyl 5-nitro-2-furoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-400-7
RTECSNumber LV2012500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only