missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-n-propyl-1H-pyrazole-3-carboxylate, 98%

CAS: 92945-27-2 | C9H14N2O2 | 182.223 g/mol

$199.48 - $766.14

Chemical Identifiers

CAS 92945-27-2
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.223
MDL Number MFCD02093950
InChI Key SUALHSUMUQQLJP-UHFFFAOYSA-N
Synonym ethyl 3-n-propylpyrazole-5-carboxylate, ethyl 3-propyl-1h-pyrazole-5-carboxylate, ethyl 5-n-propyl-1h-pyrazole-3-carboxylate, ethyl 5-propylpyrazole-3-carboxylate, 3-propyl-1h-pyrazole-5-carboxylic acid ethyl ester, 5-propyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl 5-propyl-2h-pyrazole-3-carboxylate, 3-n-propylpyrazole-5-carboxylic acid ethyl ester, ethyl 3-propylpyrazole-5-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-propyl-, ethyl ester
PubChem CID 7365260
IUPAC Name ethyl 5-propyl-1H-pyrazole-3-carboxylate
SMILES CCCC1=CC(=NN1)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1768706
View Documents
Thermo Scientific Chemicals
L1768706
5 g
Each for $199.48
Only null left
 
AAL1768714
View Documents
Thermo Scientific Chemicals
L1768714
25 g
Each for $766.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

92945-27-2
182.223
SUALHSUMUQQLJP-UHFFFAOYSA-N
7365260
CCCC1=CC(=NN1)C(=O)OCC
C9H14N2O2
MFCD02093950
ethyl 3-n-propylpyrazole-5-carboxylate, ethyl 3-propyl-1h-pyrazole-5-carboxylate, ethyl 5-n-propyl-1h-pyrazole-3-carboxylate, ethyl 5-propylpyrazole-3-carboxylate, 3-propyl-1h-pyrazole-5-carboxylic acid ethyl ester, 5-propyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl 5-propyl-2h-pyrazole-3-carboxylate, 3-n-propylpyrazole-5-carboxylic acid ethyl ester, ethyl 3-propylpyrazole-5-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-propyl-, ethyl ester
ethyl 5-propyl-1H-pyrazole-3-carboxylate

Specifications

92945-27-2
C9H14N2O2
5 g
ethyl 3-n-propylpyrazole-5-carboxylate, ethyl 3-propyl-1h-pyrazole-5-carboxylate, ethyl 5-n-propyl-1h-pyrazole-3-carboxylate, ethyl 5-propylpyrazole-3-carboxylate, 3-propyl-1h-pyrazole-5-carboxylic acid ethyl ester, 5-propyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl 5-propyl-2h-pyrazole-3-carboxylate, 3-n-propylpyrazole-5-carboxylic acid ethyl ester, ethyl 3-propylpyrazole-5-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-propyl-, ethyl ester
CCCC1=CC(=NN1)C(=O)OCC
182.223
182.22
Ethyl 5-n-propyl-1H-pyrazole-3-carboxylate
47°C to 51°C
MFCD02093950
7634535
SUALHSUMUQQLJP-UHFFFAOYSA-N
ethyl 5-propyl-1H-pyrazole-3-carboxylate
7365260
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only