missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-n-propyl-1H-pyrazole-3-carboxylate, 98%

Catalog No. AAL1768706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1768706 5 g
AAL1768714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1768706 Supplier Thermo Scientific Chemicals Supplier No. L1768706

CAS: 92945-27-2 | C9H14N2O2 | 182.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92945-27-2
Molecular Formula C9H14N2O2
Molecular Weight (g/mol) 182.223
MDL Number MFCD02093950
InChI Key SUALHSUMUQQLJP-UHFFFAOYSA-N
Synonym ethyl 3-n-propylpyrazole-5-carboxylate, ethyl 3-propyl-1h-pyrazole-5-carboxylate, ethyl 5-n-propyl-1h-pyrazole-3-carboxylate, ethyl 5-propylpyrazole-3-carboxylate, 3-propyl-1h-pyrazole-5-carboxylic acid ethyl ester, 5-propyl-2h-pyrazole-3-carboxylic acid ethyl ester, ethyl 5-propyl-2h-pyrazole-3-carboxylate, 3-n-propylpyrazole-5-carboxylic acid ethyl ester, ethyl 3-propylpyrazole-5-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-propyl-, ethyl ester
PubChem CID 7365260
IUPAC Name ethyl 5-propyl-1H-pyrazole-3-carboxylate
SMILES CCCC1=CC(=NN1)C(=O)OCC

Specifications

Melting Point 47°C to 51°C
Quantity 5 g
Beilstein 7634535
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material Ethyl 5-n-propyl-1H-pyrazole-3-carboxylate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only