missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-methoxyindole-2-carboxylate, 98%

Catalog No. AAL1759606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1759606 5 g
AAL1759603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1759606 Supplier Thermo Scientific Chemicals Supplier No. L1759606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 4792-58-9 | C12H13NO3 | 219.24 g/mol

It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.

Chemical Identifiers

CAS 4792-58-9
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00047163
InChI Key NPIUAXNFAUGNHP-UHFFFAOYSA-N
PubChem CID 20926
IUPAC Name ethyl 5-methoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OC

Specifications

Melting Point 154°C to 157°C
Quantity 5 g
Beilstein 178104
Synonym ethyl 5-methoxyindole-2-carboxylate, 5-methoxy-1h-indole-2-carboxylic acid ethyl ester, ethyl5-methoxyindole-2-carboxylate, ethyl 5-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylic acid, 5-methoxy-, ethyl ester, unii-d63u367gnz, 5-methoxyindole-2-carboxylic acid ethyl ester, methoxy-5 indole carboxylate d'ethyle-2, ethyl 5-methoxyl-1h-indole-2-carboxylate, indole-2-carboxylic acid, 5-methoxy-, ethyl ester
Solubility Information Insoluble in water.
Formula Weight 219.24
Percent Purity 98%
Chemical Name or Material Ethyl 5-methoxyindole-2-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only