CAS RN 4792-58-9
IUPAC Name:
ethyl 5-methoxy-1H-indole-2-carboxylate
Synonyms:
ethyl 5-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol):
219.24
Molecular Formula:
C12H13NO3
InChi Key:
NPIUAXNFAUGNHP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC2=CC(OC)=CC=C2N1
1
–
1
of
1
results
1
Promotions Available
Ethyl 5-methoxyindole-2-carboxylate, 98%
Ethyl 5-methoxyindole-2-carboxylate, 98%
CAS: 4792-58-9 Molecular Formula: C12H13NO3 Molecular Weight (g/mol): 219.24 MDL Number: MFCD00047163 InChI Key: NPIUAXNFAUGNHP-UHFFFAOYSA-N PubChem CID: 20926 IUPAC Name: ethyl 5-methoxy-1H-indole-2-carboxylate SMILES: CCOC(=O)C1=CC2=C(N1)C=CC(=C2)OC