missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-Chlorothiophene-2-carboxylate 98.0+%, TCI America™
$50.05 - $163.64
Chemical Identifiers
| CAS | 5751-82-6 |
|---|---|
| Molecular Formula | C7H7ClO2S |
| Molecular Weight (g/mol) | 190.641 |
| MDL Number | MFCD00051732 |
| InChI Key | BMOKMXXTOZFEIZ-UHFFFAOYSA-N |
| Synonym | 5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate |
| PubChem CID | 521808 |
| IUPAC Name | ethyl 5-chlorothiophene-2-carboxylate |
| SMILES | CCOC(=O)C1=CC=C(S1)Cl |
Chemical Identifiers
| 5751-82-6 | |
| 190.641 | |
| BMOKMXXTOZFEIZ-UHFFFAOYSA-N | |
| 521808 | |
| CCOC(=O)C1=CC=C(S1)Cl |
| C7H7ClO2S | |
| MFCD00051732 | |
| 5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate | |
| ethyl 5-chlorothiophene-2-carboxylate |
Specifications
| 5751-82-6 | |
| 62°C | |
| MFCD00051732 | |
| 5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate | |
| CCOC(=O)C1=CC=C(S1)Cl | |
| 190.641 | |
| 190.64 | |
| Liquid |
| Yellow | |
| C7H7ClO2S | |
| 1 g | |
| BMOKMXXTOZFEIZ-UHFFFAOYSA-N | |
| ethyl 5-chlorothiophene-2-carboxylate | |
| 521808 | |
| ≥98.0% (GC) | |
| Ethyl 5-Chlorothiophene-2-carboxylate |
Safety and Handling
TSCA : No