accessibility menu, dialog, popup

UserName

CAS RN 5751-82-6

CH3OOSCl

CAS RN 5751-82-6

IUPAC Name: ethyl 5-chlorothiophene-2-carboxylate
Synonyms: ethyl 5-chlorothiophene-2-carboxylate
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: BMOKMXXTOZFEIZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Cl)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results