missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Chlorothiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. E11121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11121G 1 g
E11125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11121G Supplier TCI America Supplier No. E11121G
Only null left

Chemical Identifiers

CAS 5751-82-6
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00051732
InChI Key BMOKMXXTOZFEIZ-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid ethyl ester, 5-chlorothiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid, 5-chloro-, ethyl ester, ethyl 5-chlorothiophene-2-carboxate, ethyl 5-chloro-2-thiophenecarboxylate, pubchem16133, acmc-209znt, ksc494s8t, ethyl5-chlorothiophene-2-carboxlate, ethyl 5-chloranylthiophene-2-carboxylate
PubChem CID 521808
IUPAC Name ethyl 5-chlorothiophene-2-carboxylate
SMILES CCOC(=O)C1=CC=C(S1)Cl

Specifications

Color Yellow
Boiling Point 62°C
Quantity 1 g
Formula Weight 190.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5-Chlorothiophene-2-carboxylate