missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Bromoindole-2-carboxylate 98.0+%, TCI America™

Catalog No. E06985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06985G 5 g
E06981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06985G Supplier TCI America Supplier No. E06985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16732-70-0
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD00022701
InChI Key LWRLKENDQISGEU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid ethyl ester, ethyl 5-bromoindole-2-carboxylate, 2-ethoxycarbonyl-5-bromo-indole, 5-bromo-2-indolecarboxylic acid ethyl ester, 5-bromo-1h-indole-2-carboxylic acid ethyl ester, 5-bromoindole-2-carboxylicacidethylester, 1h-indole-2-carboxylic acid, 5-bromo-, ethyl ester, 5-bromo-2-carbethoxyindole, ethyl-5-brom-1h-indol-2-carboxylat, zlchem 465
PubChem CID 259091
IUPAC Name ethyl 5-bromo-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(Br)=CC=C2N1

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 5 g
Formula Weight 268.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Bromoindole-2-carboxylate