accessibility menu, dialog, popup

UserName

H3COOHNBr

CAS RN 16732-70-0

IUPAC Name: ethyl 5-bromo-1H-indole-2-carboxylate
Synonyms: ethyl 5-bromo-1H-indole-2-carboxylate
Molecular Weight (g/mol): 268.11
Molecular Formula: C11H10BrNO2
InChi Key: LWRLKENDQISGEU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(Br)=CC=C2N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)