missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromoindole-2-carboxylate, 97%

Catalog No. AAH6679703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6679703 1 g
AAH6679706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6679703 Supplier Thermo Scientific Chemicals Supplier No. H6679703

CAS: 16732-70-0 | C11H10BrNO2 | 268.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16732-70-0
Molecular Formula C11H10BrNO2
Molecular Weight (g/mol) 268.11
MDL Number MFCD00022701
InChI Key LWRLKENDQISGEU-UHFFFAOYSA-N
Synonym 5-bromoindole-2-carboxylic acid ethyl ester, ethyl 5-bromoindole-2-carboxylate, 2-ethoxycarbonyl-5-bromo-indole, 5-bromo-2-indolecarboxylic acid ethyl ester, 5-bromo-1h-indole-2-carboxylic acid ethyl ester, 5-bromoindole-2-carboxylicacidethylester, 1h-indole-2-carboxylic acid, 5-bromo-, ethyl ester, 5-bromo-2-carbethoxyindole, ethyl-5-brom-1h-indol-2-carboxylat, zlchem 465
PubChem CID 259091
IUPAC Name ethyl 5-bromo-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(Br)=CC=C2N1

Specifications

Melting Point 163°C to 167°C
Quantity 1 g
Beilstein 170256
Formula Weight 268.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 5-bromoindole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only