missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-aminopyrazine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH66918MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66918MD 250 mg
AAH6691803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66918MD Supplier Thermo Scientific Chemicals Supplier No. H66918MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54013-06-8 | C7H9N3O2 | 167.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54013-06-8
Molecular Formula C7H9N3O2
Molecular Weight (g/mol) 167.17
MDL Number MFCD08437665
InChI Key BLPDHDYKCBRILY-UHFFFAOYSA-N
PubChem CID 22271413
IUPAC Name ethyl 5-aminopyrazine-2-carboxylate
SMILES CCOC(=O)C1=CN=C(N)C=N1

Specifications

Quantity 250 mg
Beilstein 609566
Synonym ethyl 5-amino-2-pyrazinecarboxylate, 5-amino-pyrazine-2-carboxylic acid ethyl ester, ethyl5-amino-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 5-amino-, ethyl ester, pubchem7868, ksc599g2h, ethyl5-aminopyrazine-2-carboxylate, 5-aminopyrazine-2-carboxylic acid ethyl ester, 2-pyrazinecarboxylicacid, 5-amino-, ethyl ester, 2-pyrazinecarboxylic acid, 5-amino-, ethyl ester
Formula Weight 167.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 5-aminopyrazine-2-carboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only