accessibility menu, dialog, popup

UserName

H3COONNNH2

CAS RN 54013-06-8

IUPAC Name: ethyl 5-aminopyrazine-2-carboxylate
Synonyms: ethyl 5-aminopyrazine-2-carboxylate
Molecular Weight (g/mol): 167.17
Molecular Formula: C7H9N3O2
InChi Key: BLPDHDYKCBRILY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results