missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-Aminobenzofuran-2-carboxylate 98.0+%, TCI America™
$121.16 - $367.22
Chemical Identifiers
| CAS | 174775-48-5 |
|---|---|
| Molecular Formula | C11H11NO3 |
| Molecular Weight (g/mol) | 205.21 |
| MDL Number | MFCD08275091 |
| InChI Key | YFFLLDHEEWSHQG-UHFFFAOYSA-N |
| Synonym | ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f |
| PubChem CID | 9837138 |
| IUPAC Name | ethyl 5-amino-1-benzofuran-2-carboxylate |
| SMILES | CCOC(=O)C1=CC2=CC(N)=CC=C2O1 |
Chemical Identifiers
| 174775-48-5 | |
| 205.21 | |
| YFFLLDHEEWSHQG-UHFFFAOYSA-N | |
| 9837138 | |
| CCOC(=O)C1=CC2=CC(N)=CC=C2O1 |
| C11H11NO3 | |
| MFCD08275091 | |
| ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f | |
| ethyl 5-amino-1-benzofuran-2-carboxylate |
Specifications
| 174775-48-5 | |
| C11H11NO3 | |
| 1 g | |
| YFFLLDHEEWSHQG-UHFFFAOYSA-N | |
| ethyl 5-amino-1-benzofuran-2-carboxylate | |
| 9837138 | |
| ≥98.0% (GC,T) | |
| Ethyl 5-Aminobenzofuran-2-carboxylate |
| 64°C | |
| MFCD08275091 | |
| ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f | |
| CCOC(=O)C1=CC2=CC(N)=CC=C2O1 | |
| 205.21 | |
| 205.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No