accessibility menu, dialog, popup

UserName

H3COOONH2

CAS RN 174775-48-5

IUPAC Name: ethyl 5-amino-1-benzofuran-2-carboxylate
Synonyms: ethyl 5-amino-1-benzofuran-2-carboxylate
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: YFFLLDHEEWSHQG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(N)=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results