missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Aminobenzofuran-2-carboxylate 98.0+%, TCI America™

Catalog No. E11385G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11385G 5 g
E11381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11385G Supplier TCI America Supplier No. E11385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174775-48-5
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD08275091
InChI Key YFFLLDHEEWSHQG-UHFFFAOYSA-N
Synonym ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f
PubChem CID 9837138
IUPAC Name ethyl 5-amino-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(N)=CC=C2O1

Specifications

Melting Point 64°C
Quantity 5 g
Formula Weight 205.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Aminobenzofuran-2-carboxylate