missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7129261
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
E09805G 5 g
E098025G 25 g
2 options

Catalog No. E09805G

Supplier: TCI America E09805G

Only null left

Chemical Identifiers

CAS 58168-20-0
Molecular Formula C12H14N2O4S
Molecular Weight (g/mol) 282.314
MDL Number MFCD00832845
InChI Key FLAGIUJSXKJCOB-UHFFFAOYSA-N
Synonym 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylic Acid Ethyl Ester
PubChem CID 2741370
IUPAC Name ethyl 5-amino-4-cyano-3-(2-ethoxy-2-oxoethyl)thiophene-2-carboxylate
SMILES CCOC(=O)CC1=C(SC(=C1C#N)N)C(=O)OCC

Specifications

Melting Point 139°C
Color Yellow
Quantity 5 g
Formula Weight 282.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5-Amino-4-cyano-3-(2-ethoxycarbonylmethyl)thiophene-2-carboxylate