missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate, 97+%

Catalog No. AAH2607906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2607906 5 g
AAH2607914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2607906 Supplier Thermo Scientific Chemicals Supplier No. H2607906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23286-70-6 | C7H11N3O2 | 169.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23286-70-6
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.184
MDL Number MFCD01210846
InChI Key WOCMIZZYXHVSPS-UHFFFAOYSA-N
Synonym ethyl 5-amino-3-methyl-1h-pyrazole-4-carboxylate, 5-amino-3-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-3-methylpyrazole-4-carboxylate, ethyl 3-amino-5-methyl-2h-pyrazole-4-carboxylate, acmc-1co96, 5-amino-3-methyl-1h-pyrazole-4-carboxylicacidethylester, ethyl 3-amino-5-methylpyrazole-4-carboxylate, 3-amino-5-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-methyl-5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, pyrazole-4-carboxylic acid, 3-amino-5-methyl-, ethyl ester
PubChem CID 683559
IUPAC Name ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1N)C

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only