accessibility menu, dialog, popup

UserName

CAS RN 23286-70-6

IUPAC Name: ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
Synonyms: ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 169.18
Molecular Formula: C7H11N3O2
InChi Key: WOCMIZZYXHVSPS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)NN=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results