missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate, 98%

Catalog No. p-7081525
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1153406 5 g
AAA1153414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1153406 Supplier Thermo Scientific Chemicals Supplier No. A1153406
Only null left

CAS: 31037-02-2 | C7H11N3O2 | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31037-02-2
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.18
MDL Number MFCD00051652
InChI Key MEUSJJFWVKBUFP-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
PubChem CID 271299
IUPAC Name ethyl 5-amino-1-methylpyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(C)N=C1

Specifications

Melting Point 97°C to 102°C
Quantity 5 g
Beilstein 131200
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only