missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5,6-Dichloronicotinate 98.0+%, TCI America™

Catalog No. p-7127488
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27725G 5 g
D277225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27725G Supplier TCI America Supplier No. D27725G

Chemical Identifiers

CAS 401566-69-6
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.05
MDL Number MFCD03093627
InChI Key WGQSPCCCBKQNEV-UHFFFAOYSA-N
Synonym 5,6-Dichloronicotinic Acid Ethyl Ester, 5,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester, Ethyl 5,6-Dichloropyridine-3-carboxylate
PubChem CID 16762519
IUPAC Name ethyl 5,6-dichloropyridine-3-carboxylate
SMILES CCOC(=O)C1=CC(Cl)=C(Cl)N=C1

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 5 g
Formula Weight 220.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 5,6-Dichloronicotinate