missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(Trifluoromethyl)phenylacetate 98.0+%, TCI America™

Catalog No. p-7129364
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11521G 1 g
E11525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11521G Supplier TCI America Supplier No. E11521G

Chemical Identifiers

CAS 721-63-1
Molecular Formula C11H11F3O2
Molecular Weight (g/mol) 232.20
MDL Number MFCD07779351
InChI Key BDVKGYOFECBKDX-UHFFFAOYSA-N
Synonym ethyl 2-4-trifluoromethyl phenyl acetate, ethyl 4-trifluoromethyl phenylacetate, 4-trifluoromethyl benzeneacetic acid ethyl ester, 4-trifluoromethyl-phenyl-acetic acid ethyl ester, benzeneacetic acid, 4-trifluoromethyl-, ethyl ester, ethyl4-trifluoromethyl phenylacetate, ethyl 4-trifluoromethylphenylacetate, ethyl 4-trifluoromethyl-phenyl acetate, ethyl 4-trifluoromethyl phenyl acetate
PubChem CID 14134124
IUPAC Name ethyl 2-[4-(trifluoromethyl)phenyl]acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 232.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 4-(Trifluoromethyl)phenylacetate