missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Pyridylacetate 98.0+%, TCI America™

Catalog No. P13995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13995G 5 g
P139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13995G Supplier TCI America Supplier No. P13995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54401-85-3
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00051830
InChI Key QVLJLWHOILVHJJ-UHFFFAOYSA-N
Synonym ethyl 4-pyridylacetate, ethyl 2-pyridin-4-yl acetate, ethyl pyridine-4-acetate, ethyl pyridin-4-ylacetate, 4-pyridineacetic acid ethyl ester, 4-pyridineacetic acid, ethyl ester, ethyl 2-4-pyridyl acetate, 4-pyridylacetic acid ethyl ester, ethyl 4-pyridineacetate, ethyl-4-pyridylacetate
PubChem CID 736321
IUPAC Name ethyl 2-pyridin-4-ylacetate
SMILES CCOC(=O)CC1=CC=NC=C1

Specifications

Boiling Point 128°C
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Pyridylacetate