missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Oxocyclohexanecarboxylate 98.0+%, TCI America™

Catalog No. p-7125042
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15305G 5 g
C153025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15305G Supplier TCI America Supplier No. C15305G

Chemical Identifiers

CAS 17159-79-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00013285
InChI Key ZXYAWONOWHSQRU-UHFFFAOYSA-N
Synonym ethyl 4-oxocyclohexanecarboxylate, ethyl 4-cyclohexanonecarboxylate, 4-ethoxycarbonyl cyclohexanone, ethyl cyclohexanone-4-carboxylate, ethyl-4-oxo-cyclohexane carboxylate, 4-oxo-cyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-oxo-, ethyl ester, ethyl4-oxocyclohexanecarboxylate, 4-cyclohexanonecarboxylic acid ethyl ester, ethyl 4-oxocyclohexane carboxylate
PubChem CID 317638
IUPAC Name ethyl 4-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(=O)CC1

Specifications

Color Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Oxocyclohexanecarboxylate