accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 17159-79-4

IUPAC Name: ethyl 4-oxocyclohexane-1-carboxylate
Synonyms: ethyl 4-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: ZXYAWONOWHSQRU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1CCC(=O)CC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)