missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Nitrophenylacetate 98.0+%, TCI America™

Catalog No. p-7133690
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02285G 5 g
N022825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02285G Supplier TCI America Supplier No. N02285G
Only null left

Chemical Identifiers

CAS 5445-26-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00017046
InChI Key DWDRNKYLWMKWTH-UHFFFAOYSA-N
Synonym ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester
PubChem CID 79517
IUPAC Name ethyl 2-(4-nitrophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 196°C
Quantity 5 g
Formula Weight 209.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Nitrophenylacetate