missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitrophenylacetate, 98%

Catalog No. p-7081219
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1110106 5 g
AAA1110114 25 g
AAA1110122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1110106 Supplier Thermo Scientific Chemicals Supplier No. A1110106

CAS: 5445-26-1 | C10H11NO4 | 209.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5445-26-1
Molecular Formula C10H11NO4
Molecular Weight (g/mol) 209.20
MDL Number MFCD00017046
InChI Key DWDRNKYLWMKWTH-UHFFFAOYSA-N
Synonym ethyl 4-nitrophenylacetate, ethyl 2-4-nitrophenyl acetate, ethyl p-nitrophenylacetate, ethyl 4-nitrobenzeneacetate, ethyl 4-nitrophenyl acetate, 4-nitrophenylacetic acid ethyl ester, benzeneacetic acid, 4-nitro-, ethyl ester, ethyl-4-nitrophenylacetate, acetic acid, p-nitrophenyl-, ethyl ester, 4-nitro-phenyl-acetic acid ethyl ester
PubChem CID 79517
IUPAC Name ethyl 2-(4-nitrophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 62°C to 66°C
Quantity 5 g
Beilstein 2053241
Formula Weight 209.2
Percent Purity 98%
Chemical Name or Material Ethyl 4-nitrophenylacetate
EINECSNumber 226-646-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only