missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitroindole-2-carboxylate, 97%

Catalog No. p-7054977
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63440MD 250 mg
AAH6344003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63440MD Supplier Thermo Scientific Chemicals Supplier No. H63440MD
Only null left

CAS: 4993-93-5 | C11H10N2O4 | 234.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4993-93-5
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD06795512
InChI Key LXUABEANNNKOLF-UHFFFAOYSA-N
Synonym ethyl 4-nitroindole-2-carboxylate, 4-nitro-indole-2-carboxylic acid ethyl ester, 4-nitro-1h-indole-2-carboxylic acid ethyl ester, ethyl-4-nitroindole-2-carboxylate, ethyl 4-nitro-2-indolecarboxylate, 1h-indole-2-carboxylic acid, 4-nitro-, ethyl ester
PubChem CID 283173
IUPAC Name ethyl 4-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC=C2[N+](=O)[O-]

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 234.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-nitroindole-2-carboxylate
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only