missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitrobenzoate, 98%

Catalog No. AAA1339836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339836 500 g
AAA1339814 25 g
AAA1339822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1339836 Supplier Thermo Scientific Chemicals Supplier No. A1339836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-77-4 | C9H9NO4 | 195.174 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-77-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.174
MDL Number MFCD00007351
InChI Key PHWSCBWNPZDYRI-UHFFFAOYSA-N
Synonym ethyl p-nitrobenzoate, benzoic acid, 4-nitro-, ethyl ester, 4-nitrobenzoic acid ethyl ester, ethyl nitrobenzoate, para ester, p-nitrobenzoic acid, ethyl ester, 4-ethoxycarbonyl nitrobenzene, benzoic acid, p-nitro-, ethyl ester, ethyl-p-nitrobenzoate, unii-1och6y8nar, ethyl para-nitrobenzoate
PubChem CID 7457
IUPAC Name ethyl 4-nitrobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 55°C to 58°C
Quantity 500 g
Beilstein 1912879
Merck Index 14,3832
Formula Weight 195.18
Percent Purity 98%
Chemical Name or Material Ethyl 4-nitrobenzoate
EINECSNumber 202-786-2
RTECSNumber DH5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only