missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (4-Methylphenoxy)acetate 97.0+%, TCI America™
$60.40 - $305.33
Chemical Identifiers
| CAS | 67028-40-4 |
|---|---|
| Molecular Formula | C11H14O3 |
| Molecular Weight (g/mol) | 194.23 |
| MDL Number | MFCD00209535 |
| InChI Key | UMNOIMVMNARUSB-UHFFFAOYSA-N |
| Synonym | (4-Methylphenoxy)acetic Acid Ethyl Ester |
| PubChem CID | 4047281 |
| IUPAC Name | ethyl 2-(4-methylphenoxy)acetate |
| SMILES | CCOC(=O)COC1=CC=C(C=C1)C |
Chemical Identifiers
| 67028-40-4 | |
| 194.23 | |
| UMNOIMVMNARUSB-UHFFFAOYSA-N | |
| 4047281 | |
| CCOC(=O)COC1=CC=C(C=C1)C |
| C11H14O3 | |
| MFCD00209535 | |
| (4-Methylphenoxy)acetic Acid Ethyl Ester | |
| ethyl 2-(4-methylphenoxy)acetate |
Specifications
| 67028-40-4 | |
| 152°C | |
| MFCD00209535 | |
| (4-Methylphenoxy)acetic Acid Ethyl Ester | |
| CCOC(=O)COC1=CC=C(C=C1)C | |
| 194.23 | |
| 194.23 | |
| Liquid |
| Yellow | |
| C11H14O3 | |
| 5 g | |
| UMNOIMVMNARUSB-UHFFFAOYSA-N | |
| ethyl 2-(4-methylphenoxy)acetate | |
| 4047281 | |
| ≥97.0% (GC) | |
| Ethyl (4-Methylphenoxy)acetate |
Safety and Handling
EINECSNumber : (3)-0571
TSCA : Yes