missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (4-Methylphenoxy)acetate 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 152°C |
| Quantity | 5 g |
| Formula Weight | 194.23 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl (4-Methylphenoxy)acetate |
Safety and Handling
| EINECSNumber | (3)-0571 |
| TSCA | Yes |