missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Iodobenzoate 98.0+%, TCI America™

Catalog No. I036925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I036925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I036925G Supplier TCI America Supplier No. I036925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51934-41-9
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00017344
InChI Key YCBJOQUNPLTBGG-UHFFFAOYSA-N
Synonym p-iodobenzoic acid ethyl ester, 4-iodobenzoic acid ethyl ester, ethyl p-iodobenzoate, ethyl-4-iodobenzoate, benzoicacid,4-iodo-,ethylester, 4-iodo-benzoic acid ethyl ester, benzoic acid, 4-iodo-, ethyl ester, ethyl4-iodobenzoate, ethyl-4 iodobenzoate, ethyl,4-iodobenzoate
PubChem CID 142891
IUPAC Name ethyl 4-iodobenzoate
SMILES CCOC(=O)C1=CC=C(I)C=C1

Specifications

Boiling Point 283°C
Quantity 25 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4-Iodobenzoate