missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxyquinoline-3-carboxylate 98.0+%, TCI America™

Catalog No. E11421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11421G 1 g
E11425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11421G Supplier TCI America Supplier No. E11421G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26892-90-0
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.224
MDL Number MFCD00173406
InChI Key YBEOYBKKSWUSBR-UHFFFAOYSA-N
Synonym 4-Hydroxyquinoline-3-carboxylic Acid Ethyl Ester
PubChem CID 220876
IUPAC Name ethyl 4-oxo-1H-quinoline-3-carboxylate
SMILES CCOC(=O)C1=CNC2=CC=CC=C2C1=O

Specifications

Melting Point 271°C
Quantity 1 g
Formula Weight 217.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Hydroxyquinoline-3-carboxylate