accessibility menu, dialog, popup

UserName

CAS RN 26892-90-0

IUPAC Name: ethyl 4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: ethyl 4-oxo-1H-quinoline-3-carboxylate
Molecular Weight (g/mol): 217.22
Molecular Formula: C12H11NO3
InChi Key: YBEOYBKKSWUSBR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results