missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxyphenylacetate 98.0+%, TCI America™

Catalog No. p-7129294
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10345G 5 g
E103425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10345G Supplier TCI America Supplier No. E10345G
Only null left

Chemical Identifiers

CAS 17138-28-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016491
InChI Key HYUPPKVFCGIMDB-UHFFFAOYSA-N
Synonym ethyl 4-hydroxyphenylacetate, ethyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-, ethyl ester, ethyl 4-hydroxyphenyl acetate, p-hydroxyphenyl acetic acid ethyl ester, ethyl p-hydroxyphenyl acetate, ethyl-4-hydroxyphenylacetate, pubchem12861, acetic acid, p-hydroxyphenyl-, ethyl ester
PubChem CID 28310
IUPAC Name ethyl 2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(O)C=C1

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 4-Hydroxyphenylacetate