missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxyphenylacetate, 98%

Catalog No. AAB2183218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2183218 50 g
AAB2183209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2183218 Supplier Thermo Scientific Chemicals Supplier No. B2183218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17138-28-2 | C10H12O3 | 180.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17138-28-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00016491
InChI Key HYUPPKVFCGIMDB-UHFFFAOYSA-N
Synonym ethyl 4-hydroxyphenylacetate, ethyl 2-4-hydroxyphenyl acetate, 4-hydroxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-, ethyl ester, ethyl 4-hydroxyphenyl acetate, p-hydroxyphenyl acetic acid ethyl ester, ethyl p-hydroxyphenyl acetate, ethyl-4-hydroxyphenylacetate, pubchem12861, acetic acid, p-hydroxyphenyl-, ethyl ester
PubChem CID 28310
IUPAC Name ethyl 2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(O)C=C1

Specifications

Melting Point 36°C to 38°C
Boiling Point 187°C to 189°C (20 mmHg)
Packaging Glass bottle
Quantity 50 g
Formula Weight 180.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-hydroxyphenylacetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-197-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only