missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxymandelate, 98%

Catalog No. p-7044241
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0541303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0541303 Supplier Thermo Scientific Chemicals Supplier No. L0541303

CAS: 68758-68-9 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68758-68-9
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00020180
InChI Key VLOUFSKXRCPIQR-UHFFFAOYNA-N
Synonym ethyl 2-hydroxy-2-4-hydroxyphenyl acetate, ethyl 4-hydroxymandelate, ethyl 4-hydroxy-dl-mandelate, ethyl alpha,4-dihydroxyphenylacetate, 4-hydroxymandelic acid ethyl ester, benzeneacetic acid, .alpha.,4-dihydroxy-, ethyl ester, benzeneacetic acid, alpha,4-dihydroxy-, ethyl ester, ethyl hydroxy 4-hydroxyphenyl acetate, ethyl hydroxy 4-hydroxyphenyl acetate #, 4-2-ethoxy-1-hydroxy-2-oxoethyl phenol
PubChem CID 111357
IUPAC Name ethyl 2-hydroxy-2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)C(O)C1=CC=C(O)C=C1

Specifications

Melting Point 128°C to 132°C
Quantity 1 g
Beilstein 3132065
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Ethyl 4-hydroxymandelate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only