missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. h0211500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
H0211500G 500 g
H021125G 25 g
2 options

Catalog No. H0211500G

Supplier: TCI America H0211500G

Only null left

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 117°C
Color White
Boiling Point 178°C
Quantity 500 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH2190000
TSCA Yes