missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-Hydroxybenzoate 99.0+%, TCI America™
Supplier: TCI America E088425G
Specifications
| Ethyl 4-Hydroxybenzoate [for Biochemical Research] | |
| 117°C | |
| 178°C | |
| MFCD00002353 | |
| ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e | |
| CCOC(=O)C1=CC=C(C=C1)O | |
| 166.176 | |
| CHEBI:86616 | |
| ≥99.0% (GC,T) |
| 120-47-8 | |
| White | |
| C9H10O3 | |
| 25 g | |
| NUVBSKCKDOMJSU-UHFFFAOYSA-N | |
| ethyl 4-hydroxybenzoate | |
| 8434 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 120-47-8 | |
| 166.176 | |
| NUVBSKCKDOMJSU-UHFFFAOYSA-N | |
| 8434 | |
| ethyl 4-hydroxybenzoate |
| C9H10O3 | |
| MFCD00002353 | |
| ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e | |
| CHEBI:86616 | |
| CCOC(=O)C1=CC=C(C=C1)O |
Safety and Handling
EINECSNumber : (3)-1585
RTECSNumber : DH2190000
TSCA : Yes