missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxybenzoate, 99%

CAS: 120-47-8 | C9H10O3 | 166.18 g/mol

$72.85 - $72.85

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC150251000
SDS View Documents
Thermo Scientific Chemicals
150251000
100 g
Each for $72.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

120-47-8
166.18
NUVBSKCKDOMJSU-UHFFFAOYSA-N
8434
ethyl 4-hydroxybenzoate
C9H10O3
MFCD00002353
ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
CHEBI:86616
CCOC(=O)C1=CC=C(C=C1)O

Specifications

120-47-8
White
Authentic
Plastic bottle
HOC6H4CO2C2H5
100 g
15, 3890
Solubility in water: 1.7g/L (20°C). Other solubilities: freely soluble in alcohol and ether,soluble in carbon disulfide,slightly soluble in petroleum ether,chloroform
CCOC(=O)C1=CC=C(C=C1)O
166.18
CHEBI:86616
99%
Ethyl 4-hydroxybenzoate
115°C to 118°C
297°C to 298°C
98.5% min. (GC)
C9H10O3
MFCD00002353
10, 159
ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
NUVBSKCKDOMJSU-UHFFFAOYSA-N
ethyl 4-hydroxybenzoate
8434
166.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-399-4

SDS
Documents

Documents

RUO – Research Use Only