missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Ethyl 4-hydroxybenzoate

Catalog No. 111988500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111988500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111988500G Supplier Sigma Aldrich Supplier No. 111988500G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 4-hydroxybenzoate

Specifications

CAS 120-47-8
Melting Point 114°C to 117°C (lit.)
Boiling Point 297°C to 298°C (lit.)
Molecular Formula C9H10O3
Linear Formula HOC6H4CO2C2H5
MDL Number MFCD00002353
Quantity 500 g
Synonym Ethylparaben
Molecular Weight (g/mol) 166.17
Percent Purity 99%