missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxy-3-nitrobenzoate, 98%

Catalog No. AAL0645106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0645106 5 g
AAL0645114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0645106 Supplier Thermo Scientific Chemicals Supplier No. L0645106

CAS: 19013-10-6 | C9H9NO5 | 211.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19013-10-6
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.173
MDL Number MFCD00016999
InChI Key FBHJNBWUGONVNS-UHFFFAOYSA-N
Synonym benzoic acid, 4-hydroxy-3-nitro-, ethyl ester, 4-hydroxy-3-nitrobenzoic acid ethyl ester, acmc-20akyj, ethyl4-hydroxy-3-nitrobenzoate, ethyl 3-nitro-4-hydroxybenzoate, ethyl 4-hydroxy-3-nitro benzoate, 3-nitro-4-hydroxybenzoic acid ethyl ester, 4-hydroxy-3-nitro-benzoic acid ethyl ester
PubChem CID 87895
IUPAC Name ethyl 4-hydroxy-3-nitrobenzoate
SMILES CCOC(=O)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 69°C to 71°C
Quantity 5 g
Beilstein 2121319
Formula Weight 211.18
Percent Purity 98%
Chemical Name or Material Ethyl 4-hydroxy-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-751-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only