Learn More
Ethyl 4-formylpyrrole-2-carboxylate, 96%
CAS: 7126-57-0 | C8H9NO3 | 167.164 g/mol
Supplier: Thermo Scientific Chemicals H6477603
Description
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.
Solubility
Miscible with ethyl acetate.
Notes
Incompatible with steel and aluminum.
Specifications
| Ethyl 4-formylpyrrole-2-carboxylate | |
| 101°C to 105°C | |
| MFCD04122505 | |
| 473842 | |
| Miscible with ethyl acetate. | |
| CCOC(=O)C1=CC(=CN1)C=O | |
| 167.164 | |
| 167.16 |
| 7126-57-0 | |
| C8H9NO3 | |
| 1 g | |
| ethyl 4-formylpyrrole-2-carboxylate, 4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-formyl-, ethyl ester, acmc-20akyk, pubchem14845, ethyl4-formyl-1h-pyrrole-2-carboxylate, ethyl 4-methanoyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-, ethyl ester | |
| WVSAWXIWWNJTAV-UHFFFAOYSA-N | |
| ethyl 4-formyl-1H-pyrrole-2-carboxylate | |
| 321486 | |
| 96% |
Chemical Identifiers
| 7126-57-0 | |
| 167.164 | |
| WVSAWXIWWNJTAV-UHFFFAOYSA-N | |
| 321486 | |
| CCOC(=O)C1=CC(=CN1)C=O |
| C8H9NO3 | |
| MFCD04122505 | |
| ethyl 4-formylpyrrole-2-carboxylate, 4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-formyl-, ethyl ester, acmc-20akyk, pubchem14845, ethyl4-formyl-1h-pyrrole-2-carboxylate, ethyl 4-methanoyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-, ethyl ester | |
| ethyl 4-formyl-1H-pyrrole-2-carboxylate |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only