missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-formylpyrrole-2-carboxylate, 96%

Catalog No. p-7041702
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64776MD 250 mg
AAH6477603 1 g
AAH6477606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64776MD Supplier Thermo Scientific Chemicals Supplier No. H64776MD
Only null left

CAS: 7126-57-0 | C8H9NO3 | 167.164 g/mol

Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

Solubility
Miscible with ethyl acetate.

Notes
Incompatible with steel and aluminum.

Chemical Identifiers

CAS 7126-57-0
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD04122505
InChI Key WVSAWXIWWNJTAV-UHFFFAOYSA-N
Synonym ethyl 4-formylpyrrole-2-carboxylate, 4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester, 1h-pyrrole-2-carboxylic acid, 4-formyl-, ethyl ester, acmc-20akyk, pubchem14845, ethyl4-formyl-1h-pyrrole-2-carboxylate, ethyl 4-methanoyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylicacid, 4-formyl-, ethyl ester
PubChem CID 321486
IUPAC Name ethyl 4-formyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC(=CN1)C=O

Specifications

Melting Point 101°C to 105°C
Quantity 250 mg
Beilstein 473842
Solubility Information Miscible with ethyl acetate.
Formula Weight 167.16
Percent Purity 96%
Chemical Name or Material Ethyl 4-formylpyrrole-2-carboxylate
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only